ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Metabotropic Glutamate Receptor 7/MGLUR7 protein to be purified and analysed in vitro. 1 * 100 µG
  ANTIA317310-100
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Metabotropic Glutamate Receptor 7/MGLUR7 protein to be purified and analysed in vitro. 1 * 100 µG
  ANTIA317310-100