ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Prostaglandin F2 alpha Receptor/PTGFR protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325283-50
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Prostaglandin F2 alpha Receptor/PTGFR protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325283-50
Supplier:  ANTIBODIES.COM