Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Metabotropic Glutamate Receptor 2/MGLUR2 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA317296-100
Supplier: Antibodies.com