Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Omicron B.1.1.529 Variant) containing the following muta tions: G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H. The h 1 * 50 µG
Catalog # ANTIA318058-50
Dodavatel: Antibodies.com