ContactUsLinkComponent
This Thioredoxin Peroxidase / Periredoxin 4 Assay Kit is a quantitative assay kit for measuring Thioredoxin Peroxidase / Peroxiredox in 4 activity in serum, plasma, animal tissues, cells, and bacteria. 1 * 48 Tests
Catalog # ANTIA319744-48
Supplier:  ANTIBODIES.COM
New Product
This Thioredoxin Peroxidase / Periredoxin 4 Assay Kit is a quantitative assay kit for measuring Thioredoxin Peroxidase / Peroxiredox in 4 activity in serum, plasma, animal tissues, cells, and bacteria. 1 * 48 Tests
Catalog # ANTIA319744-48
Supplier:  ANTIBODIES.COM