ContactUsLinkComponent
Human MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection antib ody, Streptavidin-AP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched ant 1 * 1.920 Tests
Catalog # ANTIA325346-2096
Supplier:  ANTIBODIES.COM
New Product
Human MDC Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an biotin-labelled monoclonal detection antib ody, Streptavidin-AP, recombinant human CCL22 ELISA standard, and standard reconstitution buffer A5. This matched ant 1 * 1.920 Tests
Catalog # ANTIA325346-2096
Supplier:  ANTIBODIES.COM