ContactUsLinkComponent
Mouse Proteinase-3 Anti-Neutrophil Cytoplasmic Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Proteinase-3 Anti-Neutrophil Cytoplasmic Antibody in serum, plasm 1 * 96 Tests
Catalog # ANTIA303541-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Proteinase-3 Anti-Neutrophil Cytoplasmic Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Proteinase-3 Anti-Neutrophil Cytoplasmic Antibody in serum, plasm 1 * 96 Tests
Catalog # ANTIA303541-96
Supplier:  ANTIBODIES.COM