ContactUsLinkComponent
Human Epstein Barr Virus Early Antigen IgM ELISA Kit is a trapping Enzyme-Linked Immunosorbent Assay designed for the in vitro quali tative determination of human Epstein Barr Virus Early Antigen IgM in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA303094-96
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus Early Antigen IgM ELISA Kit is a trapping Enzyme-Linked Immunosorbent Assay designed for the in vitro quali tative determination of human Epstein Barr Virus Early Antigen IgM in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA303094-96
Supplier:  ANTIBODIES.COM