ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Somatostatin Receptor 2 protein to be purified and analysed in vitro. 1 * 100 µG
  ANTIA318484-100
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Somatostatin Receptor 2 protein to be purified and analysed in vitro. 1 * 100 µG
  ANTIA318484-100