Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Adenosine A3 Receptor/A3AR protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325086-10
Supplier: Antibodies.com