ContactUsLinkComponent
A heterodimer of Integrin alpha 2 (Tyr30-Thr1132) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal h uman Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318046-100
Supplier:  Antibodies.com
New Product
A heterodimer of Integrin alpha 2 (Tyr30-Thr1132) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal h uman Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318046-100
Supplier:  Antibodies.com