ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Melatonin Receptor 1A/MTNR1A protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325229-100
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Melatonin Receptor 1A/MTNR1A protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325229-100
Supplier:  Antibodies.com