ContactUsLinkComponent
Bovine Plasminogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of bovine Plasminogen in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA326185-48
Supplier:  ANTIBODIES.COM
New Product
Bovine Plasminogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of bovine Plasminogen in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA326185-48
Supplier:  ANTIBODIES.COM