ContactUsLinkComponent
Monkey IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA325374-696
Supplier:  Antibodies.com
New Product
Monkey IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA325374-696
Supplier:  Antibodies.com