ContactUsLinkComponent
Rat NFkB p105 / p50 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat NFkB p105 / p50 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2877-48
Supplier:  ANTIBODIES.COM
New Product
Rat NFkB p105 / p50 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat NFkB p105 / p50 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2877-48
Supplier:  ANTIBODIES.COM