ContactUsLinkComponent
Monkey IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Catalog # ANTIA325375-696
Supplier:  Antibodies.com
New Product
Monkey IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human IL-10 ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Catalog # ANTIA325375-696
Supplier:  Antibodies.com