ContactUsLinkComponent
Porcine Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Lipocalin-2/NGAL in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3171-48
Supplier:  Antibodies.com
New Product
Porcine Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Lipocalin-2/NGAL in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3171-48
Supplier:  Antibodies.com