ContactUsLinkComponent
Human Sonic Hedgehog ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Sonic Hedgehog in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2867-48
Supplier:  ANTIBODIES.COM
New Product
Human Sonic Hedgehog ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Sonic Hedgehog in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2867-48
Supplier:  ANTIBODIES.COM