ContactUsLinkComponent
Human Kallikrein 3/KLK3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Kallikrein 3/KLK3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA3087-48
Supplier:  ANTIBODIES.COM
New Product
Human Kallikrein 3/KLK3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Kallikrein 3/KLK3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA3087-48
Supplier:  ANTIBODIES.COM