ContactUsLinkComponent
Mouse TRAP/CD40L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse TRAP/CD40L in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3054-48
Supplier:  ANTIBODIES.COM
New Product
Mouse TRAP/CD40L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse TRAP/CD40L in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3054-48
Supplier:  ANTIBODIES.COM