ContactUsLinkComponent
Mouse Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA303540-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA303540-96
Supplier:  ANTIBODIES.COM