ContactUsLinkComponent
Bovine TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant bovine TNF alpha ELISA standard, and standard reconstitution buffer. This match 1 * 576 Tests
Bovine TNF alpha Matched Antibody Pair Kit
Catalog # ANTIA325324-696
Supplier:  Antibodies.com
New Product
CAS Number:  
Bovine TNF alpha Matched Antibody Pair Kit
Catalog # ANTIA325324-696
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Frequently Bought Together