ContactUsLinkComponent
Thyroxine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Thyroxine in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3162-48
Supplier:  ANTIBODIES.COM
New Product
Thyroxine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Thyroxine in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3162-48
Supplier:  ANTIBODIES.COM