Human Epstein-Barr Virus Early Antigen IgG Antibody ELISA Kit is a 90 minute Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Epstein-Barr Virus Early Antigen IgG antibody in serum, plasma, and n 1 * 96 Tests
Catalog # ANTIA314417-96
Supplier: ANTIBODIES.COM