ContactUsLinkComponent
Porcine Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Interferon gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA3242-48
Supplier:  ANTIBODIES.COM
New Product
Porcine Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of porcine Interferon gamma in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA3242-48
Supplier:  ANTIBODIES.COM