ContactUsLinkComponent
This Phenylalanine Ammonia Lyase Activity Assay Kit is a quantitative assay kit for measuring Phenylalanine Ammonia Lyase activity i n plant tissues. 1 * 96 Tests
Catalog # ANTIA319714-96
Supplier:  Antibodies.com
New Product
This Phenylalanine Ammonia Lyase Activity Assay Kit is a quantitative assay kit for measuring Phenylalanine Ammonia Lyase activity i n plant tissues. 1 * 96 Tests
Catalog # ANTIA319714-96
Supplier:  Antibodies.com