Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Mu Opioid Receptor protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325242-100
Dodavatel: Antibodies.com