ContactUsLinkComponent
Rat Procollagen I C-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Procollagen I C-Terminal Propeptide in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3169-48
Supplier:  ANTIBODIES.COM
New Product
Rat Procollagen I C-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Procollagen I C-Terminal Propeptide in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3169-48
Supplier:  ANTIBODIES.COM