Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Thromboxane A2 Receptor/TBXA2R protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325304-50
Supplier: ANTIBODIES.COM