Horse POMC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f horse POMC in serum, plasma, saliva or other biological fluids. 1 * 96 Tests
Katalogové číslo ANTIA326186-96
Dodavatel: ANTIBODIES.COM