This Mitochondrial Malate Dehydrogenase Activity Assay Kit is a quantitative assay kit for measuring Mitochondrial Malate Dehydrogen ase activity in animal and plant tissues, cells, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319705-48
Supplier: ANTIBODIES.COM