ContactUsLinkComponent
Human Anti-Human CD3 Epsilon Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitati ve determination of human Anti-Human CD3 Epsilon Antibody in serum, plasma, cell culture supernatant, cell or tissue 1 * 96 Tests
Catalog # ANTIA326371-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-Human CD3 Epsilon Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitati ve determination of human Anti-Human CD3 Epsilon Antibody in serum, plasma, cell culture supernatant, cell or tissue 1 * 96 Tests
Catalog # ANTIA326371-96
Supplier:  ANTIBODIES.COM