ContactUsLinkComponent
Porcine Cystatin C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Cystatin C in serum, plasma, urine or other biological fluids. 1 * 96 Tests
  ANTIA3172-96
New Product
Porcine Cystatin C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Cystatin C in serum, plasma, urine or other biological fluids. 1 * 96 Tests
  ANTIA3172-96