ContactUsLinkComponent
Human CXCL13 ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro quantitative determination of human CXCL13 i n cerebrospinal fluid. 1 * 96 Tests
Catalog # ANTIA324300-96
Supplier:  ANTIBODIES.COM
New Product
Human CXCL13 ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro quantitative determination of human CXCL13 i n cerebrospinal fluid. 1 * 96 Tests
Catalog # ANTIA324300-96
Supplier:  ANTIBODIES.COM