ContactUsLinkComponent
This Na+/K+-ATPase Activity Assay Kit is a quantitative assay kit for measuring Na+/K+-A TPase activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319707-48
Supplier:  ANTIBODIES.COM
New Product
This Na+/K+-ATPase Activity Assay Kit is a quantitative assay kit for measuring Na+/K+-A TPase activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319707-48
Supplier:  ANTIBODIES.COM