ContactUsLinkComponent
Horse Cardiac Troponin I ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitativ e determination of horse Cardiac Troponin I in serum, plasma, cell culture supernatant, cell or tissue lysate, and ot 1 * 96 Tests
Catalog # ANTIA326507-96
Supplier:  ANTIBODIES.COM
New Product
Horse Cardiac Troponin I ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitativ e determination of horse Cardiac Troponin I in serum, plasma, cell culture supernatant, cell or tissue lysate, and ot 1 * 96 Tests
Catalog # ANTIA326507-96
Supplier:  ANTIBODIES.COM