ContactUsLinkComponent
Human CXCL1/GRO alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human CXCL1/GRO alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA326137-48
Supplier:  ANTIBODIES.COM
New Product
Human CXCL1/GRO alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human CXCL1/GRO alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA326137-48
Supplier:  ANTIBODIES.COM