ContactUsLinkComponent
Human Perinuclear Anti-Neutrophil Cytoplasmic Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed f or the in vitro quantitative determination of human Perinuclear Anti-Neutrophil Cytoplasmic Antibody in serum, plasma 1 * 96 Tests
Catalog # ANTIA302952-96
Supplier:  ANTIBODIES.COM
New Product
Human Perinuclear Anti-Neutrophil Cytoplasmic Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed f or the in vitro quantitative determination of human Perinuclear Anti-Neutrophil Cytoplasmic Antibody in serum, plasma 1 * 96 Tests
Catalog # ANTIA302952-96
Supplier:  ANTIBODIES.COM