ContactUsLinkComponent
Monkey Fibrinogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey Fibrinogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sample 1 * 96 Tests
Catalog # ANTIA326563-96
Supplier:  ANTIBODIES.COM
New Product
Monkey Fibrinogen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey Fibrinogen in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sample 1 * 96 Tests
Catalog # ANTIA326563-96
Supplier:  ANTIBODIES.COM