ContactUsLinkComponent
Mouse Renin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Renin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sample volume 1 * 96 Tests
Catalog # ANTIA323201-96
Supplier:  Antibodies.com
New Product
Mouse Renin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Renin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sample volume 1 * 96 Tests
Catalog # ANTIA323201-96
Supplier:  Antibodies.com