ContactUsLinkComponent
This 6-Phosphogluconate Dehydrogenase Activity Assay Kit is a quantitative assay kit for measuring 6-Phosphogluconate Dehydrogenase activity in serum, plasma, animal and plant tissues, cells, cell culture supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319653-96
Supplier:  ANTIBODIES.COM
New Product
This 6-Phosphogluconate Dehydrogenase Activity Assay Kit is a quantitative assay kit for measuring 6-Phosphogluconate Dehydrogenase activity in serum, plasma, animal and plant tissues, cells, cell culture supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319653-96
Supplier:  ANTIBODIES.COM