ContactUsLinkComponent
Human Mycoplasma pneumoniae IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determin ation of human Mycoplasma pneumoniae IgM in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324331-96
Supplier:  ANTIBODIES.COM
New Product
Human Mycoplasma pneumoniae IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determin ation of human Mycoplasma pneumoniae IgM in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324331-96
Supplier:  ANTIBODIES.COM