ContactUsLinkComponent
Rat Oncostatin M/OSM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Oncostatin M/OSM in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3445-96
Supplier:  ANTIBODIES.COM
New Product
Rat Oncostatin M/OSM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Oncostatin M/OSM in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3445-96
Supplier:  ANTIBODIES.COM