ContactUsLinkComponent
Mouse Myostatin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Myostatin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Katalogové číslo ANTIA3342-96
Dodavatel:  ANTIBODIES.COM
New Product
Mouse Myostatin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Myostatin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Katalogové číslo ANTIA3342-96
Dodavatel:  ANTIBODIES.COM