ContactUsLinkComponent
Human Cystatin SN/CST1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Cystatin SN/CST1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Human Cystatin SN/CST1 ELISA Kit
Catalog # ANTIA4996-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Human Cystatin SN/CST1 ELISA Kit
Catalog # ANTIA4996-48
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Frequently Bought Together