ContactUsLinkComponent
Bovine Procollagen II N-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of bovine Procollagen II N-Terminal Propeptide in serum, plasma, tissue homogenates o 1 * 96 Tests
Catalog # ANTIA4441-96
Supplier:  ANTIBODIES.COM
New Product
Bovine Procollagen II N-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of bovine Procollagen II N-Terminal Propeptide in serum, plasma, tissue homogenates o 1 * 96 Tests
Catalog # ANTIA4441-96
Supplier:  ANTIBODIES.COM