ContactUsLinkComponent
Human VILIP3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human VILIP3 in tissue homogenates, cerebrospinal fluid or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4899-96
Supplier:  ANTIBODIES.COM
New Product
Human VILIP3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human VILIP3 in tissue homogenates, cerebrospinal fluid or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4899-96
Supplier:  ANTIBODIES.COM