ContactUsLinkComponent
Nitrofurazone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Nitrofurazone i n honey, tissue, casing, liver. 1 * 50 Tests
Catalog # ANTIA327233-50
Supplier:  Antibodies.com
New Product
Nitrofurazone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Nitrofurazone i n honey, tissue, casing, liver. 1 * 50 Tests
Catalog # ANTIA327233-50
Supplier:  Antibodies.com