ContactUsLinkComponent
Human TCIRG1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TCIRG1 in serum or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5412-96
Supplier:  ANTIBODIES.COM
New Product
Human TCIRG1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TCIRG1 in serum or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5412-96
Supplier:  ANTIBODIES.COM