ContactUsLinkComponent
Mouse IL-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse IL-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA3579-48
Supplier:  Antibodies.com
New Product
Mouse IL-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse IL-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA3579-48
Supplier:  Antibodies.com